Learn More
ABclonal Technology pAbPC1 Rabbit pAb
SDP
Supplier:  ABclonal Technology A148721000UL
Encompass_Preferred
This gene encodes a poly(A) binding protein. The protein shuttles between the nucleus and cytoplasm and binds to the 3 poly(A) tail of eukaryotic messenger RNAs via RNA-recognition motifs. The binding of this protein to poly(A) promotes ribosome recruitment and translation initiation, it is also required for poly(A) shortening which is the first step in mRNA decay. The gene is part of a small gene family including three protein-coding genes and several pseudogenes.
Catalog No. 50-001-05294