Learn More
Aviva Systems Biology Corp PABPC1L2A Antibody - middle region (ARP40286_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40286_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PABPC1L2A. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human PABPC1L2A. Alternate names: RBM32A, PABPC1L2B, RP11-493K23.3ApplicationWB. Species reactivity: Cow, Guinea Pig, Horse, Human, Pig, Rabbit, Rat
Catalog No. 50-014-57376