Learn More
Aviva Systems Biology Corp PABPC4 Antibody - middle region (ARP40495_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40495_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PABPC4. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human PABPC4. Alternate names: APP-1, APP1, PABP4, iPABPApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-57486