Learn More
Apexbio Technology LLC PAC-1 315183-21-2 250mg
SDP
Catalog No. 5000090167
Encompass_Preferred
PAC-1 (CAS 315183-21-2) is a small molecule activator of procaspase-3 promoting its conversion to active caspase-3 through time-dependent cleavage This direct activation of caspase-3 leads to rapid induction of apoptosis in cancer cell lines The compound s pro-apoptotic effect has been shown to correlate with intracellular procaspase-3 levels and its ability to induce cell death displays IC50 values ranging from 0 003 to 1 41 M in early-stage colon cancer isolates compared to higher IC50 values in adjacent normal tissues PAC-1 serves as a valuable tool for investigating apoptosis mechanisms and caspase-3-associated anticancer strategies
Catalog No. 50-000-90167 Supplier Apexbio Technology LLC Supplier No. A8177250
PAC-1 (CAS 315183-21-2) is a small molecule activator of procaspase-3 promoting its conversion to active caspase-3 through time-dependent cleavage This direct activation of caspase-3 leads to rapid induction of apoptosis in cancer cell lines The compound s pro-apoptotic effect has been shown to correlate with intracellular procaspase-3 levels and its ability to induce cell death displays IC50 values ranging from 0 003 to 1 41 M in early-stage colon cancer isolates compared to higher IC50 values in adjacent normal tissues PAC-1 serves as a valuable tool for investigating apoptosis mechanisms and caspase-3-associated anticancer strategies