Learn More
Abcam PAC-1, caspase-3 activator, 10MG
Catalog No. 502706348
Encompass_Preferred
MW 392.5 Da, Purity >95%. Potent caspase-3 activator (IC₅₀ = 220 nM). Also activates caspase-7 (IC₅₀ = 4.5 μM). Proapoptotic in vitro. Anticancer activity in vivo. Orally active.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6348 Supplier Abcam Supplier No. AB14207410MG
MW 392.5 Da, Purity >95%. Potent caspase-3 activator (IC₅₀ = 220 nM). Also activates caspase-7 (IC₅₀ = 4.5 μM). Proapoptotic in vitro. Anticancer activity in vivo. Orally active.
The product is subject to the following: Abcam Restricted Use Statement