Learn More
MSC Pac Steel Strapping, 1/2 x 0.02 in, Oscillated, 3087 ft
Catalog No. 5000796187
Encompass
This regular duty steel strapping is supplied on an oscillated coil, designed for various packaging applications. It offers consistent performance in manual, pneumatic, and automatic strapping heads.
  • Conforms to ASTM specification D3953
  • Consistent in width, gauge, and finish
  • Usable in manual, pneumatic, and automatic strapping heads
  • Painted, waxed, and edge-treated to the highest quality
Catalog No. 50-007-96187 Supplier MSC Supplier No. 01700657
This regular duty steel strapping is supplied on an oscillated coil, designed for various packaging applications. It offers consistent performance in manual, pneumatic, and automatic strapping heads.
  • Conforms to ASTM specification D3953
  • Consistent in width, gauge, and finish
  • Usable in manual, pneumatic, and automatic strapping heads
  • Painted, waxed, and edge-treated to the highest quality