Learn More
MSC Pac Strapping Feedwheel Tensioner, 3/4 to 1-1/4 in, 1 ea.
Catalog No. 5000796232
Encompass
A feedwheel tensioner designed for manual strapping applications, suitable for use with steel strapping.
  • Heavy-duty construction
  • Manual power source
  • Compatible with 3/4 in to 1-1/4 in steel strapping
  • Accommodates strapping thickness from 0.0312 in up to 0.044 in
  • Manufactured in Taiwan
Catalog No. 50-007-96232 Supplier MSC Supplier No. 01700897
A feedwheel tensioner designed for manual strapping applications, suitable for use with steel strapping.
  • Heavy-duty construction
  • Manual power source
  • Compatible with 3/4 in to 1-1/4 in steel strapping
  • Accommodates strapping thickness from 0.0312 in up to 0.044 in
  • Manufactured in Taiwan