Learn More
MSC Pac Strapping Sealer, 1/2 in, Front Action, Heavy-Duty, 1 ea
Catalog No. 5000770194
Encompass
This Pac Strapping sealer is a manual, heavy-duty tool designed for front-action sealing. It is specifically engineered for use with 1/2 in PET and HSS-4A strapping, providing an open function for efficient operation.
  • Sealer, front action
  • For use with 1/2 in PET and HSS-4A strapping
  • For strapping width: 1/2 in
  • Tool function: open
  • Tool power source: manual
  • Tool strength: heavy-duty
Catalog No. 50-007-70194 Supplier MSC Supplier No. 01700913
This Pac Strapping sealer is a manual, heavy-duty tool designed for front-action sealing. It is specifically engineered for use with 1/2 in PET and HSS-4A strapping, providing an open function for efficient operation.
  • Sealer, front action
  • For use with 1/2 in PET and HSS-4A strapping
  • For strapping width: 1/2 in
  • Tool function: open
  • Tool power source: manual
  • Tool strength: heavy-duty