Learn More
MSC Pac Strapping Steel Strapping, 1/2 in., 0.02 in., 865 ft.
Catalog No. 5000675705
Encompass
This steel strapping, measuring 1/2 in. wide and 0.02 in. thick with a length of 865 ft., is supplied as a ribbon coil. It offers a breaking strength of 800 lb, making it suitable for demanding packaging and bundling tasks.
  • Available in various regular duty steel strapping sizes
  • Conforms to ASTM specification D3953
  • Consistent in width, gauge, and finish
  • Compatible with manual, pneumatic, and automatic strapping heads
  • Painted, waxed, and edge-treated for high quality
  • Offered in full pallets and portable coils
Catalog No. 50-006-75705 Supplier MSC Supplier No. 81549057
This steel strapping, measuring 1/2 in. wide and 0.02 in. thick with a length of 865 ft., is supplied as a ribbon coil. It offers a breaking strength of 800 lb, making it suitable for demanding packaging and bundling tasks.
  • Available in various regular duty steel strapping sizes
  • Conforms to ASTM specification D3953
  • Consistent in width, gauge, and finish
  • Compatible with manual, pneumatic, and automatic strapping heads
  • Painted, waxed, and edge-treated for high quality
  • Offered in full pallets and portable coils