Learn More
MSC Pac Strapping Steel Strapping, 3/4 in, 0.02 in, 1 Roll
Catalog No. 5000770219
Encompass
This regular duty steel strapping is designed for secure packaging and bundling. It features consistent width, gauge, and finish, making it suitable for various strapping applications. Each roll contains 2058 ft of strapping.
  • Conforms to ASTM D3953
  • Consistent in width, gauge, and finish
  • Usable with manual, pneumatic, and automatic strapping heads
  • Painted, waxed, and edge-treated for quality
  • Available in full pallets and portable coils
Catalog No. 50-007-70219 Supplier MSC Supplier No. 01700699
This regular duty steel strapping is designed for secure packaging and bundling. It features consistent width, gauge, and finish, making it suitable for various strapping applications. Each roll contains 2058 ft of strapping.
  • Conforms to ASTM D3953
  • Consistent in width, gauge, and finish
  • Usable with manual, pneumatic, and automatic strapping heads
  • Painted, waxed, and edge-treated for quality
  • Available in full pallets and portable coils