Learn More
MSC Pac Strapping Steel Strapping, 5/8 in, 0.023 in, 1 Roll
Catalog No. 5000796234
Encompass
This regular duty steel strapping is designed for reliable use in manual, pneumatic, and automatic strapping heads. It is manufactured with consistent width, gauge, and finish, conforming to ASTM specification D3953.
  • Consistent width, gauge, and finish
  • Can be used in manual, pneumatic, and automatic strapping heads
  • Painted, waxed, and edge-treated for high quality
  • Conforms to ASTM specification D3953
  • Available in full pallets and portable coils
Catalog No. 50-007-96234 Supplier MSC Supplier No. 01700681
This regular duty steel strapping is designed for reliable use in manual, pneumatic, and automatic strapping heads. It is manufactured with consistent width, gauge, and finish, conforming to ASTM specification D3953.
  • Consistent width, gauge, and finish
  • Can be used in manual, pneumatic, and automatic strapping heads
  • Painted, waxed, and edge-treated for high quality
  • Conforms to ASTM specification D3953
  • Available in full pallets and portable coils