Learn More
MSC Pac Strapping Steel Strapping Kit, 1/2 in, Portable, 1 Kit
Catalog No. 5000792808
Encompass
This portable steel strapping kit is designed for demanding applications requiring high strength and low elongation, providing a secure solution for heavy-duty packaging and bundling. It includes all the necessary tools and materials for efficient and reliable steel strapping operations.
  • Two coils of 200 ft, 1/2 in x 0.020 in steel strapping
  • Open steel seals (500)
  • Tensioner
  • Double notch crimper
  • Steel strap cutter
  • Heavy-duty reusable dispenser
  • Reusable steel carrying case
  • Instructions
Catalog No. 50-007-92808 Supplier MSC Supplier No. 67208108
This portable steel strapping kit is designed for demanding applications requiring high strength and low elongation, providing a secure solution for heavy-duty packaging and bundling. It includes all the necessary tools and materials for efficient and reliable steel strapping operations.
  • Two coils of 200 ft, 1/2 in x 0.020 in steel strapping
  • Open steel seals (500)
  • Tensioner
  • Double notch crimper
  • Steel strap cutter
  • Heavy-duty reusable dispenser
  • Reusable steel carrying case
  • Instructions