Learn More
Medchemexpress LLC PACAP (1-27) | 127317-03-7 | 99.3% | 3147.66 | 5 MG
SDP
Catalog No. 5000426401
Encompass_Preferred
PACAP (1-27), human, ovine, rat (PACAP 1-27) is the N-terminal fragment of PACAP-38. It is a potent PACAP receptor agonist with IC50s of 3 nM for rat PAC1, 2 nM for rat VPAC1, and 5 nM for human VPAC2. This product is for research use only.
  • Potent PACAP receptor agonist
  • N-terminal fragment of PACAP-38
  • Displaces radioligand binding in PAC-receptors on AR4-2J cells
  • Stimulates IP3 and cAMP formation in AR4-2J cells
  • Reduces total pulmonary resistance in guinea-pigs
  • Inhibitory effect on pulmonary resistance similar to salbutamol
  • Increases heart rate less than salbutamol
Catalog No. 50-004-26401 Supplier Medchemexpress LLC Supplier No. HYP01765MG
PACAP (1-27), human, ovine, rat (PACAP 1-27) is the N-terminal fragment of PACAP-38. It is a potent PACAP receptor agonist with IC50s of 3 nM for rat PAC1, 2 nM for rat VPAC1, and 5 nM for human VPAC2. This product is for research use only.
  • Potent PACAP receptor agonist
  • N-terminal fragment of PACAP-38
  • Displaces radioligand binding in PAC-receptors on AR4-2J cells
  • Stimulates IP3 and cAMP formation in AR4-2J cells
  • Reduces total pulmonary resistance in guinea-pigs
  • Inhibitory effect on pulmonary resistance similar to salbutamol
  • Increases heart rate less than salbutamol