Learn More
Medchemexpress LLC HY-P0176A 1mg Medchemexpress, PACAP (1-27), human, ovine, rat (TFA) CAS: Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP0176A1MG
Encompass_Preferred
Medchemexpress, HY-P0176A 1mg PACAP (1-27), human, ovine, rat (TFA) CAS: PACAP (1-27), human, ovine, rat TFA (PACAP 1-27 TFA) is the N-terminal fragment of PACAP-38, and is a potent PACAP receptor antagonist with IC50s of 3 nM, 2 nM and 5 nM for rat PAC1, rat VPAC1 and human VPAC2, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0518