Learn More
Selleck Chemical LLC PACAP 1-38-1mg
SDP
Supplier:  Selleck Chemical LLC S84151MG
Encompass_Preferred
PACAP 1-38 (Pituitary Adenylate Cyclase Activating Polypeptide 38) is a highly potent PACAP receptor agonist (Kd = 100 pM). It stimulates adenylate cyclase and phagocytosis.
Catalog No. 50-239-0058