Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC PACAP (1-38), amide, Biotin-labeled | 98.3% | 4760.55 | 1 MG
SDP

Catalog No. 5000423465
Encompass_Preferred

PACAP (1-38), amide, Biotin-labeled is a biologically active peptide, provided as a white to off-white solid, and is available for research use only. It features a molecular weight of 4760.55 and a purity of 98.29%.

  • Biologically active peptide
  • White to off-white solid appearance
  • For research use only
  • Molecular weight: 4760.55
  • Purity: 98.29%
  • Recommended for sealed storage, away from moisture and light

Catalog No. 50-004-23465 Supplier Medchemexpress LLC Supplier No. HYP2609F1MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

PACAP (1-38), amide, Biotin-labeled is a biologically active peptide, provided as a white to off-white solid, and is available for research use only. It features a molecular weight of 4760.55 and a purity of 98.29%.

  • Biologically active peptide
  • White to off-white solid appearance
  • For research use only
  • Molecular weight: 4760.55
  • Purity: 98.29%
  • Recommended for sealed storage, away from moisture and light

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.