Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC PACAP (1-38) free acid | 129405-61-4 | 99.94% | 4535.24 | 1 MG
SDP

Catalog No. 5000425380
Encompass_Preferred

PACAP (1-38) free acid is a desamido-PACAP (1-38) peptide, found in human, ovine, and rat species. It can activate the human PAC1 receptor and induces cyclic AMP (cAMP) generation in both NS-1 neuroendocrine cells and non-neuroendocrine HEK293 cells. This product promotes neuroendocrine cell differentiation in NS-1 cells with an EC50 of 10.6 nM and induces cell growth arrest in NS-1 cells.

  • Appears as a white to off-white solid
  • Purity: 99.94%
  • Molecular weight: 4535.24
  • Molecular formula: C203H330N62O54S
  • Sequence: His-Ser-Asp-Gly-Ile-Phe-Thr-Asp-Ser-Tyr-Ser-Arg-Tyr-Arg-Lys-Gln-Met-Ala-Val-Lys-Lys-Tyr-Leu-Ala-Ala-Val-Leu-Gly-Lys-Arg-Tyr-Lys-Gln-Arg-Val-Lys-Asn-Lys

Catalog No. 50-004-25380 Supplier Medchemexpress LLC Supplier No. HYP0221C1MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

PACAP (1-38) free acid is a desamido-PACAP (1-38) peptide, found in human, ovine, and rat species. It can activate the human PAC1 receptor and induces cyclic AMP (cAMP) generation in both NS-1 neuroendocrine cells and non-neuroendocrine HEK293 cells. This product promotes neuroendocrine cell differentiation in NS-1 cells with an EC50 of 10.6 nM and induces cell growth arrest in NS-1 cells.

  • Appears as a white to off-white solid
  • Purity: 99.94%
  • Molecular weight: 4535.24
  • Molecular formula: C203H330N62O54S
  • Sequence: His-Ser-Asp-Gly-Ile-Phe-Thr-Asp-Ser-Tyr-Ser-Arg-Tyr-Arg-Lys-Gln-Met-Ala-Val-Lys-Lys-Tyr-Leu-Ala-Ala-Val-Leu-Gly-Lys-Arg-Tyr-Lys-Gln-Arg-Val-Lys-Asn-Lys

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.