Learn More
Medchemexpress LLC HY-P0221 1mg Medchemexpress, PACAP (1-38), human, ovine, rat CAS:137061-48-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP02211MG
Encompass_Preferred
Medchemexpress, HY-P0221 1mg PACAP (1-38), human, ovine, rat CAS:137061-48-4 PACAP (1-38), human, ovine, rat is a neuropeptide with 38 amino acid residues. PACAP (1-38) binds to PACAP type I receptor , PACAP type II receptor VIP 1 , and PACAP type II receptor VIP 2 with IC 50 s of 4 nM, 2 nM, and 1 nM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-4057