Learn More
Medchemexpress LLC PACAP-38 (31-38), human, mouse, rat | 138764-85-9 | 98.8% | 1062.27 | 10 MG

Supplier: Medchemexpress LLC HYP184510MG
PACAP-38 (31-38), human, mouse, rat is a fragment containing the C-terminal 31-38 fragment of PACAP (1-38), human, ovine, rat. This fragment does not bind to the PACAP receptor.
- For research use only.
- Available in various quantities.
- Solid appearance, white to off-white color.
- Sequence: Tyr-Lys-Gln-Arg-Val-Lys-Asn-Lys-NH2 (YKQRVKNK-NH2).
- Shipping at room temperature in continental US.
- Storage: Sealed, away from moisture.
- Solubility in H2O: 100 mg/mL (94.14 mM; requires ultrasonic).
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.