Learn More
Bachem PACAP-38 (6-38) (human, chicken, mouse, ovine, porcine, rat) Trifluoroacetate
SDP
Supplier:  Bachem 4031158.1000
Encompass_Preferred
PACAP-38 (6-38) (human, chicken, mouse, ovine, porcine, rat) Trifluoroacetate, H-Phe-Thr-Asp-Ser-Tyr-Ser-Arg-Tyr-Arg-Lys-Gln-Met-Ala-Val-Lys-Lys-Tyr-Leu-Ala-Ala-Val-Leu-Gly-Lys-Arg-Tyr-Lys-Gln-Arg-Val-Lys-Asn-Lys-NH2, 143748-18-9, C182H300N56O45S, Mr 4024.80, 1mg. Pituitary Adenylate Cyclase Activating Polypeptide-38 (6-38) (human, chicken, mouse, ovine, porcine, rat). The PACAP selective antagonist PACAP (6-38) is a potent and selective inhibitor of PACAP-27-stimulated pituitary adenylate cyclase with a Ki values of 150 nM.
Catalog No. 50-259-645