Learn More
Selleck Chemical LLC PACAP 6-38 acetate
SDP
Catalog No. 5000002196
Encompass_Preferred
PACAP 6-38 acetate is a PACAP (pituitary adenylate cyclase-activating polypeptide) non-stimulating competitive antagonist with an IC50 value of 2 nM It also acts as a functional CARTp antagonist in vivo
Catalog No. 50-000-02196 Supplier Selleck Chemical LLC Supplier No. S84165MG
PACAP 6-38 acetate is a PACAP (pituitary adenylate cyclase-activating polypeptide) non-stimulating competitive antagonist with an IC50 value of 2 nM It also acts as a functional CARTp antagonist in vivo