Learn More
Medchemexpress LLC HY-P0220A 1mg Medchemexpress, PACAP (6-38), human, ovine, rat (TFA) CAS: Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP0220A1MG
Encompass_Preferred
Medchemexpress, HY-P0220A 1mg PACAP (6-38), human, ovine, rat (TFA) CAS: PACAP (6-38), human, ovine, rat TFA is a potent PACAP receptor antagonist with IC 50 s of 30, 600, and 40 nM for PACAP type I receptor , PACAP type II receptor VIP 1 , and PACAP type II receptor VIP 2 , respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-4056