Learn More
Medchemexpress LLC PACAP (6-38), human, ovine, rat | 143748-18-9 | 99.1% | 4024.74 | 5 MG
SDP
Catalog No. 5000425994
Encompass_Preferred
PACAP (6-38), human, ovine, rat TFA is a potent PACAP receptor antagonist with IC50s of 30 nM for the PACAP type I receptor, 600 nM for the PACAP type II receptor VIP1, and 40 nM for the PACAP type II receptor VIP2. It inhibits the increase of dopamine content and cell proliferation induced by Dexamethasone in PC12 cells.
  • Potent PACAP receptor antagonist
  • Inhibits PACAP type 1 (PAC1) receptor
  • Increases intercontraction interval and void volume in NGF-OE mice
  • Decreases baseline bladder pressure in NGF-OE mice
  • Reduces pelvic sensitivity in NGF-OE mice
  • For research use only
Catalog No. 50-004-25994 Supplier Medchemexpress LLC Supplier No. HYP02205MG
PACAP (6-38), human, ovine, rat TFA is a potent PACAP receptor antagonist with IC50s of 30 nM for the PACAP type I receptor, 600 nM for the PACAP type II receptor VIP1, and 40 nM for the PACAP type II receptor VIP2. It inhibits the increase of dopamine content and cell proliferation induced by Dexamethasone in PC12 cells.
  • Potent PACAP receptor antagonist
  • Inhibits PACAP type 1 (PAC1) receptor
  • Increases intercontraction interval and void volume in NGF-OE mice
  • Decreases baseline bladder pressure in NGF-OE mice
  • Reduces pelvic sensitivity in NGF-OE mice
  • For research use only