Learn More
CPC Scientific H-His-Ser-Asp-Gly-Ile-Phe-Thr-Asp-Ser-Tyr-Ser-Arg-Tyr-Arg-Lys-Gln-Met-Ala-Val-Lys-Lys-Tyr-Leu-Ala-Ala-Val-Leu-NH2 (trifluoroacetate salt) 0.5MG
SDP
Supplier:  CPC Scientific PACA004A
Encompass
PACAP-38 and its N-terminal fragment PACAP-27 are novel neuropeptides originally isolated from bovine hypothalamus. They are also found in humans and rats. Both PACAP-27 and PACAP-38, showing considerable homology with vasoactive intestinal polypeptide (VIP), can stimulate adenylate cyclase much more potently than VIP.SEQUENCE: H-His-Ser-Asp-Gly-Ile-Phe-Thr-Asp-Ser-Tyr-Ser-Arg-Tyr-Arg-Lys-Gln-Met-Ala-Val-Lys-Lys-Tyr-Leu-Ala-Ala-Val-Leu-NH2 (trifluoroacetate salt)ONE-LETTER SEQUENCE: HSDGIFTDSYSRYRKQMAVKKYLAAVL-NH2MOLECULAR FORMULA: C142H224N40O39S1MOLECULAR WEIGHT: 3147.65STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: [127317-03-7]SYNONYMS: Pituitary Adenylate Cyclase Activating Polypeptide-27 (human, mouse, ovine, porcine, rat), PACAP-27 (human, mouse, ovine, porcine, rat)RESEARCH AREA: HormonalREFERENCES:A. Miyata et al., BBRC, 164, 567 (1989)A. Mirata et al., Regulatory Peptides, 26, 170 (1989)SKU(s): PACA-004A, PACA-004B
Catalog No. 50-211-0058