Learn More
CPC Scientific H-Phe-Thr-Asp-Ser-Tyr-Ser-Arg-Tyr-Arg-Lys-Gln-Met-Ala-Val-Lys-Lys-Tyr-Leu-Ala-Ala-Val-Leu-NH2 (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific PACA005B
Encompass
A specific inhibitor of adenylate cyclase activation that results in clear functional discrimination between PACAP-27 receptors and PACAP-38 receptors.SEQUENCE: H-Phe-Thr-Asp-Ser-Tyr-Ser-Arg-Tyr-Arg-Lys-Gln-Met-Ala-Val-Lys-Lys-Tyr-Leu-Ala-Ala-Val-Leu-NH2 (trifluoroacetate salt)ONE-LETTER SEQUENCE: FTDSYSRYRKQMAVKKYLAAVL-NH2MOLECULAR FORMULA: C121H193N33O31S1MOLECULAR WEIGHT: 2638.2STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: [136134-68-4]SYNONYMS: RESEARCH AREA: HormonalREFERENCES:P. Robberecht et al., FEBS Letters, 286, 133 (1991)SKU(s): PACA-005A, PACA-005B
Catalog No. 50-211-0061