Learn More
Genscript Corporation PACAP-Related Peptide (PRP), human
Supplier:  Genscript Corporation RP103410.5
Encompass
GenScript's high-quality PACAP-related peptide (PRP), a 29 amino-acid region of the PACAP precursor protein, is synthesized in quantities sufficient for biological and structural studies. PRP has a distinct biological activity on the gallbladder that is similar to PACAP but opposite to that of VIP and its related peptide, PHM.
Catalog No. 50-274-804