Learn More
Cayman Chemical PACAP-related Peptide (human) (trifluoroacetate salt), Purity: ≥95%, 1 mg
SDP
Catalog No. 502933785
Encompass_Preferred
An endogenous 29-amino acid peptide that belongs to the secretin/glucagon superfamily; is expressed in normal human pancreas and adrenal gland tissue and in some VIP-producing tumors; is secreted by CHO-K1 cells that express human recombinant preproPACAP
Catalog No. 50-293-3785 Supplier Cayman Chemical Supplier No. 247721 MG
An endogenous 29-amino acid peptide that belongs to the secretin/glucagon superfamily; is expressed in normal human pancreas and adrenal gland tissue and in some VIP-producing tumors; is secreted by CHO-K1 cells that express human recombinant preproPACAP