Learn More
Cayman Chemical PACAP-related Peptide (rat) (trifluoroacetate salt), Purity: ≥95%, 1 mg
SDP
Catalog No. 502933786
Encompass_Preferred
An endogenous 29-amino acid peptide that belongs to the secretin/glucagon superfamily of peptides; expressed in rat hypothalamus as well as within the nerves of the median eminence, the anterior pituitary, bed nucleus of the stria terminalis, cerebellum, cerebral cortex, and amygdala; expressed in vaginal, uterine cervical, uterine horn, fallopian tube, and ovarian tissues from the rat female genital tract; present in extracts of male testis tissue
Catalog No. 50-293-3786 Supplier Cayman Chemical Supplier No. 247731 MG
An endogenous 29-amino acid peptide that belongs to the secretin/glucagon superfamily of peptides; expressed in rat hypothalamus as well as within the nerves of the median eminence, the anterior pituitary, bed nucleus of the stria terminalis, cerebellum, cerebral cortex, and amygdala; expressed in vaginal, uterine cervical, uterine horn, fallopian tube, and ovarian tissues from the rat female genital tract; present in extracts of male testis tissue