Learn More
PACIFIC BIOSCIENCES OF CALIFORNIA INC PacBio Iso-Seq Express 2.0 Kit, 24 Reactions, Full-Length cDNA Generation, 12 Barcoded Primers
Catalog No. NC3977090
Encompass
The Iso-Seq express 2.0 kit provides reagents and oligos for generating full-length cDNA. This kit includes 12 unique barcoded cDNA primers for multiplexing and is compatible with the Kinnex full-length RNA kit. All components are conveniently packaged in a single box, supporting 24 reactions per kit.
  • Contains reagents and oligos for generating full-length cDNA.
  • Includes 12 unique barcoded cDNA primers for multiplexing.
  • Compatible with the Kinnex full-length RNA kit.
  • Supports 24 reactions per kit.
  • Compatible with the Revio system, Vega system, and Sequel II and IIe systems.
  • Store frozen between -15°C to -25°C.
Catalog No. NC3977090 Supplier PACIFIC BIOSCIENCES OF CALIFORNIA INC Supplier No. 103071500
The Iso-Seq express 2.0 kit provides reagents and oligos for generating full-length cDNA. This kit includes 12 unique barcoded cDNA primers for multiplexing and is compatible with the Kinnex full-length RNA kit. All components are conveniently packaged in a single box, supporting 24 reactions per kit.
  • Contains reagents and oligos for generating full-length cDNA.
  • Includes 12 unique barcoded cDNA primers for multiplexing.
  • Compatible with the Kinnex full-length RNA kit.
  • Supports 24 reactions per kit.
  • Compatible with the Revio system, Vega system, and Sequel II and IIe systems.
  • Store frozen between -15°C to -25°C.