Learn More
Medchemexpress LLC HY-N0371 5mg Medchemexpress, Pachymic acid CAS:29070-92-6 Purity:>98%
SDP
Catalog No. 501933541
Encompass_Preferred
Medchemexpress, HY-N0371 5mg Pachymic acid CAS:29070-92-6 Pachymic acid is a lanostrane-type triterpenoid from P. cocos. Pachymic acid inhibits Akt and ERK signaling pathways. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3541 Supplier Medchemexpress LLC Supplier No. HYN03715MG
Medchemexpress, HY-N0371 5mg Pachymic acid CAS:29070-92-6 Pachymic acid is a lanostrane-type triterpenoid from P. cocos. Pachymic acid inhibits Akt and ERK signaling pathways. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.