Learn More
Selleck Chemical LLC Pachymic acid-E0214-5MG
SDP
Catalog No. 503137915
Encompass_Preferred
Pachymic acid(3-O-Acetyltumulosic acid) inhibits the activity of CYP2C9 with the IC50 of 21 25 M
Catalog No. 50-313-7915 Supplier Selleck Chemical LLC Supplier No. E02145MG
Add to Cart
Add to Cart
Pachymic acid(3-O-Acetyltumulosic acid) inhibits the activity of CYP2C9 with the IC50 of 21 25 M