Learn More
New England Biolabs, Inc. PacI – 1250 units
SDP
Catalog No. 50812224
Encompass
A restriction endonuclease that recognizes the sequence TTA_AT^TAA.
Catalog No. 50-812-224 Supplier New England Biolabs, Inc. Supplier No. R0547L
A restriction endonuclease that recognizes the sequence TTA_AT^TAA.