Learn More
New England Biolabs, Inc. PacI – 250 units
SDP
Catalog No. 50812225
Encompass
A restriction endonuclease that recognizes the sequence TTA_AT^TAA.
Catalog No. 50-812-225 Supplier New England Biolabs, Inc. Supplier No. R0547S
A restriction endonuclease that recognizes the sequence TTA_AT^TAA.