Learn More
PACIFIC BIOSCIENCES OF CALIFORNIA INC Pacific Biosciences Elution Buffer, 50 mL, pH 7.0–8.0
Catalog No. NC3977120
Encompass
The Elution Buffer is a clear, colorless liquid designed for use with the PacBio™ Sequel™ system. It is suitable for workflows requiring additional elution buffer and AMPure™ PB beads cleanup steps.
  • Supplied at 50 mL volume.
  • Has a pH range of 7.0–8.0.
  • Should be stored refrigerated at 2°C to 8°C.
  • Intended for research use only, not for diagnostic procedures.
  • Not classified as a hazardous substance.
Catalog No. NC3977120 Supplier PACIFIC BIOSCIENCES OF CALIFORNIA INC Supplier No. 101633500
The Elution Buffer is a clear, colorless liquid designed for use with the PacBio™ Sequel™ system. It is suitable for workflows requiring additional elution buffer and AMPure™ PB beads cleanup steps.
  • Supplied at 50 mL volume.
  • Has a pH range of 7.0–8.0.
  • Should be stored refrigerated at 2°C to 8°C.
  • Intended for research use only, not for diagnostic procedures.
  • Not classified as a hazardous substance.