Learn More
PACIFIC BIOSCIENCES OF CALIFORNIA INC PACIFIC BIOSCIENCES Nanobind HT 1 mL Blood Kit, Automated, HMW DNA Extraction
Catalog No. NC3977063
Encompass
The Nanobind HT 1 mL blood kit supports automated high-throughput (HT) high molecular weight (HMW) DNA extraction for 1 mL of whole human blood. This kit is designed for automation on specific robotic instruments, including the Thermo Fisher KingFisher and Hamilton NIMBUS Presto platforms.
  • Supports automated high-throughput high molecular weight DNA extraction for 1 mL of whole human blood.
  • Designed for use with compatible robotic automation platforms.
  • Enables sample preparation automation on four different robotic platforms.
  • Supports 96 reactions per kit.
Catalog No. NC3977063 Supplier PACIFIC BIOSCIENCES OF CALIFORNIA INC Supplier No. 102762800
The Nanobind HT 1 mL blood kit supports automated high-throughput (HT) high molecular weight (HMW) DNA extraction for 1 mL of whole human blood. This kit is designed for automation on specific robotic instruments, including the Thermo Fisher KingFisher and Hamilton NIMBUS Presto platforms.
  • Supports automated high-throughput high molecular weight DNA extraction for 1 mL of whole human blood.
  • Designed for use with compatible robotic automation platforms.
  • Enables sample preparation automation on four different robotic platforms.
  • Supports 96 reactions per kit.