Learn More
PACIFIC BIOSCIENCES OF CALIFORNIA INC PACIFIC BIOSCIENCES OF CALIFORNIA INC Barcoded M13 Primer Plate, 384-Well, 10 μM
Catalog No. NC3977138
Encompass
This 384-well Barcoded M13 primer plate contains barcoded forward and reverse primer pairs in each well. Each primer features an M13 primer sequence on the 3\' end, enabling barcode incorporation into templates using a two-step PCR strategy.
  • Includes 16 unique forward and 24 unique reverse barcodes.
  • Primer concentration is 10 μM for both forward and reverse mixes.
  • Volume of 12 μL per well.
  • Supports 384 reactions.
  • Compatible with Revio, Sequel II, and IIe systems.
  • Store frozen between -15°C to -25°C.
  • For research use only; not for diagnostic procedures.
Catalog No. NC3977138 Supplier PACIFIC BIOSCIENCES OF CALIFORNIA INC Supplier No. 102135500
This 384-well Barcoded M13 primer plate contains barcoded forward and reverse primer pairs in each well. Each primer features an M13 primer sequence on the 3\' end, enabling barcode incorporation into templates using a two-step PCR strategy.
  • Includes 16 unique forward and 24 unique reverse barcodes.
  • Primer concentration is 10 μM for both forward and reverse mixes.
  • Volume of 12 μL per well.
  • Supports 384 reactions.
  • Compatible with Revio, Sequel II, and IIe systems.
  • Store frozen between -15°C to -25°C.
  • For research use only; not for diagnostic procedures.