Learn More
PACIFIC BIOSCIENCES OF CALIFORNIA INC Pacific Biosciences Revio SPRQ Reagent Kit, 24-pack, for Revio SMRT Cells, HiFi Sequencing
Supplier:  PACIFIC BIOSCIENCES OF CALIFORNIA INC 103520200
Encompass
This kit provides all necessary reagents for sequencing 24 SMRT cells using SPRQ chemistry, enabling high-fidelity (HiFi) sequencing on the Revio system.
  • All reagents for sequencing 24 SMRT cells.
  • Utilizes SPRQ chemistry.
  • Supports HiFi sequencing.
  • Designed for the Revio system.
Catalog No. NC3977157