Learn More
AdipoGen Pacific Blue
SDP
Supplier:  AdipoGen CDXD0079M050
Encompass_Preferred
Chemical. CAS 215868-31-8. Formula C10H4F2O5. MW 242.13. Synthetic Fluorinated fluorescent dye. Increased resistance to photobleaching and higher quantum yields makes it a superior fluorescent dye. Pacific Blue dyes yield conjugates that are strongly fluorescent, even at neutral pH. Spectral data Extinction 403nm/Emmission 455nm. The amine-reactive succinimidyl ester derivative can be used to create bright blue-fluorescent bioconjugates with excitation/emission maxima ~410/455 nm that are excitable by the 405 nm spectral line of the blue diode violet laser.
Catalog No. 50-256-1256