Learn More
Biolegend Pacific Blue™ anti-human IgD
Supplier:  Biolegend 348224
Encompass_Preferred
Pacific Blue™ anti-human IgD [IA6-2]; Isotype: Mouse IgG2a, κ; Reactivity: Human; Apps: FC; Size: 100 tests
Catalog No. NC1248098