Learn More
Biolegend Pacific Blue™ anti-human Siglec-9
Supplier:  Biolegend 351508
Encompass_Preferred
Pacific Blue™ anti-human Siglec-9 [K8]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 100 tests
Catalog No. NC1654416