Learn More
Biolegend Pacific Blue™ anti-mouse/human Ki-67 [11F6]; Isotype: Rat IgG2b, κ; Reactivity: Mouse, Human; Apps: ICFC; Size: 25 ug
Supplier:  Biolegend 151223
Encompass_Preferred
Pacific Blue™ anti-mouse/human Ki-67 [11F6]; Isotype: Rat IgG2b, κ; Reactivity: Mouse, Human; Apps: ICFC; Size: 25 ug
Catalog No. 50-237-4933