Learn More
Biolegend Pacific BlueTM anti-human CD18
Catalog No. 502722365
Encompass_Preferred
Pacific Blue™ anti-human CD182 (CXCR2) [5E8/CXCR2]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 25 tests
Catalog No. 50-272-2365 Supplier Biolegend Supplier No. 320723
Pacific Blue™ anti-human CD182 (CXCR2) [5E8/CXCR2]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 25 tests