Learn More
Thermo Scientific™ Pacific Hemostasis™ Coagulation Controls
Catalog No. 23033240 Shop All Thermo Scientific Products
Change view
Click to view available options
Type:
Fibrinogen High Control
Fibrinogen Low Control
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
23-033240 Fibrinogen High Control
23-305081 Fibrinogen Low Control
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 23-033240
Add to Cart
Add to Cart
This item is not returnable. View return policy
Provide accurate, reproducible confirmation of coagulation test system performance with these coagulation controls.
Provide accurate, reproducible confirmation of coagulation test system performance with Thermo Scientific™ Pacific Hemostasis™ Coagulation Controls. Controls can be run on most coagulation systems and are formulated to reflect expected patient results.
Coagulation and Heparin control plasmas are prepared from fresh pools of human plasma.
Coagulation Control Plasmas:
  • Normal control (Level 1) produces normal PT and aPTT times
  • Abnormal controls monitor therapeutic (Level 2) and highly elevated (Level 3) PT/aPTT times 
Heparin Control Plasmas:
  • Simulate low (Level 1) and elevated (Level 2) plasma levels of Heparin
  • Use to monitor and evaluate patients on Heparin therapy
TRUSTED_SUSTAINABILITY
Specifications
Type Fibrinogen High Control
Quantity 10 x 1 mL