Learn More
Thermo Scientific™ Pacific Hemostasis™ FDP Assay
Catalog No. p-4528065
Change view
Click to view available options
Quantity:
2 x 100 mL
30 Determinations
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-262567 2 x 100 mL
23-249989 30 Determinations
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 23-262567
This item is not returnable. View return policy
Fisherbrand™ Pacific Hemostasis FDP Assay is a fast, easy-to-use latex agglutination assay for use in measuring fibrinogen degradation products (FDP) in serum and urine.
Measure fibrinogen degradation products (FDP) in serum and urine with Thermo Scientific™ Pacific Hemostasis™ FDP Assay, a fast, easy-to-use latex agglutination assay used in the diagnosis of disseminated intravascular coagulation (DIC), deep vein thrombosis (DVT), pulmonary embolism (PE), certain pregnancy disorders and myocardial infarction.
FDP is used in the diagnosis of disseminated intravascular coagulation (DIC), deep vein thrombosis (DVT) and pulmonary embolism (PE). Additionally, it is used to diagnose certain pregnancy disorders and myocardial infarction.
  • Latex sensitivity of 1ug/mL of FDP
  • No pretreatment of heparin specimen
  • No repeat testing due to false positives from heparin interference
TRUSTED_SUSTAINABILITY
Specifications
Description Glycerine Buffered Saline
Quantity 2 x 100 mL