Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ SickleScreen™ Control Set, 4 x 0.5mL
GSA_VA

Catalog No. 22251051 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
4 vials x 0.5mL bottles
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-251051 4 vials x 0.5mL bottles
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 22-251051
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Control Kit for use with Thermo Scientific SickleScreen Assay Set, 22-251050

For use with Thermo Scientific SickleScreen Assay Set.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Description SickleScreen Control Set
Includes Positive and Negative Controls; Each level -4 bottles each level x 5 ml fill, Reconstitutiom Control Kit - 2 bottles x 4 mL per vial
Quantity 4 vials x 0.5mL bottles
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.