Learn More
Thermo Scientific™ SickleScreen™ Assay Kit
GSA_VA
Catalog No. 22251050 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
1
30-Determination Kit
Type:
SickleScreen Assay Set
SickleScreen Reading Rack
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Type
22-251050 30-Determination Kit SickleScreen Assay Set
22-251053 1 SickleScreen Reading Rack
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 22-251050
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy
Screen for sickle cell disease and sickle cell trait with this assay kit.
Screen for sickle cell disease and sickle cell trait with the Fisherbrand™ Pacific Hemostasis™ SickleScreen™ Assay Kit.
Thermo Scientific SickleScreen products use a modified Nalbandian procedure that provides fast and accurate results for screening and detecting hereditary sickle cell disease and sickle cell trait. When sickle cell patients are in crisis, their Hb S becomes sickle or cresent shaped and can no longer carry oxygen. The abnormal cells often break apart and can cause damage to the spleen, kidneys, bone, eyes, lungs and central nervous system.
  • The procedure is based on the differential solubility of Hemoglobin S (Hb S) and Hemoglobin A (Hb A) in concentrated phosphate solution.
  • The 120-test kit contains phosphate buffer (4 x 125mL) and four sodium hydrosulfite vials.
  • The 30-test kit contains a bottle of phosphate buffer (125mL) with a dispensing cap and 30 pre-filled reaction vials
  • Positive and negative controls are for Hb A and Hb S.
Ordering Alerts:
The control kit (22-251-051) must be ordered separately.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
TRUSTED_SUSTAINABILITY
Specifications
Type SickleScreen Assay Set
Includes Prefilled vials with sodium hydrosulfite powder, phosphate buffer 1 x 125 mL with dispensing cap.
Quantity 30-Determination Kit