Description
- Dual injected nylon and rubber frame provides a durable frame and cushioned brow protection
- Frame is vented to allow the free flow of air to prevent fogging
- Flexible, soft nose piece and temple tips
- Lenses provide 99% protection against harmful UV rays
Specifications
Specifications
| Lens Tint | Gray |
| Frame Color | Black |
| Lens Material | Polycarbonate |
| Frame Material | Dual injected nylon/rubber |