Learn More
Med Vet International Miltex Standard Veterinary Orthopedic Kit
Catalog No. 502831835
Encompass_Preferred
The Miltex 6840 Veterinary Orthopedic instrument kit contains 18 German grade stainless steel instruments. The kit contains (1) 4.75 Wire Cutting Scissors (1) 6-3/8in. Senn Retractor (1) 5.5in. Weitlaner Retractor with 3x4 Teeth (1) 7in. Freer Elevator (1) 6.75in. Spratt Mastoid Curette, size 0 (1) 7.75in. Luace Mallet (1) 8.25in. Fomon Rasp (1) 6in. Wire Twister, Tungsten Carbide (1)Gigli-Strully Saw Handle (1) 9.5in. Kern Bone Holding Clamp (1)6.75in. Kern Bone Holding Forceps (1) 7.25in. Cannulated Pin Cutter (1) 5.5in. Liston Bone Cutting Forceps, Straight (1) 5in. Swiss Pattern Osteotome (1) Veterinary Orthopedic Storage Cassette
Catalog No. 50-283-1835 Supplier Med Vet International Supplier No. MIL6840
The Miltex 6840 Veterinary Orthopedic instrument kit contains 18 German grade stainless steel instruments. The kit contains (1) 4.75 Wire Cutting Scissors (1) 6-3/8in. Senn Retractor (1) 5.5in. Weitlaner Retractor with 3x4 Teeth (1) 7in. Freer Elevator (1) 6.75in. Spratt Mastoid Curette, size 0 (1) 7.75in. Luace Mallet (1) 8.25in. Fomon Rasp (1) 6in. Wire Twister, Tungsten Carbide (1)Gigli-Strully Saw Handle (1) 9.5in. Kern Bone Holding Clamp (1)6.75in. Kern Bone Holding Forceps (1) 7.25in. Cannulated Pin Cutter (1) 5.5in. Liston Bone Cutting Forceps, Straight (1) 5in. Swiss Pattern Osteotome (1) Veterinary Orthopedic Storage Cassette