Learn More
Chemglass Life Sciences Chemglass pack blue bonded closure and 2.0 mL glass vials, 9 mm thread, 12 x 32 mm, PTFE/red rubber septa (3 x 100)
SDP
Catalog No. 502733161
Encompass_Preferred
Convenience pack of 2.0 mL clear borosilicate glass autosampler vials with bonded blue closures and PTFE/red rubber septa. Vials include a gradation spot and printed graduations for 0.5, 1.0, and 1.5 mL; closures are bonded to reduce septa fallout and ensure reliable sealing. Supplied as three packs of 100 (300 total), these vials are suitable for chromatography autosamplers and general sample storage and handling.
  • Pack of 2.0 mL clear borosilicate glass vials.
  • 12 x 32 mm vial dimensions with 9 mm thread.
  • Blue bonded closures eliminate septa fallout.
  • PTFE/red rubber septa for chemical resistance and sealing.
  • Graduated mark spots for 0.5, 1.0, and 1.5 mL volume checks.
  • Supplied as three packs of 100 (300 total).
Catalog No. 50-273-3161 Supplier Chemglass Life Sciences Supplier No. CV400080WS
Convenience pack of 2.0 mL clear borosilicate glass autosampler vials with bonded blue closures and PTFE/red rubber septa. Vials include a gradation spot and printed graduations for 0.5, 1.0, and 1.5 mL; closures are bonded to reduce septa fallout and ensure reliable sealing. Supplied as three packs of 100 (300 total), these vials are suitable for chromatography autosamplers and general sample storage and handling.
  • Pack of 2.0 mL clear borosilicate glass vials.
  • 12 x 32 mm vial dimensions with 9 mm thread.
  • Blue bonded closures eliminate septa fallout.
  • PTFE/red rubber septa for chemical resistance and sealing.
  • Graduated mark spots for 0.5, 1.0, and 1.5 mL volume checks.
  • Supplied as three packs of 100 (300 total).