Learn More
Chemglass Life Sciences Pack, Blue Bonded Closure and Vial, Glass, 2.0mL, 9mm Thread, Clear, 12 x 32mm, PTFE/Silicone Septa with Slit
SDP
Catalog No. 5000980673
Encompass_Preferred
These convenience packs come with 100 bonded closures and vials packed seperately. Closures with bonded septa are not attached.
Catalog No. 50-009-80673 Supplier Chemglass Life Sciences Supplier No. CV53241232
These convenience packs come with 100 bonded closures and vials packed seperately. Closures with bonded septa are not attached.